Learn More
OHAUS Courier 1000 Series Shipping Scale
Catalog No. 01920197
Change view
Click to view available options
Model:
i-C12P20
i-C12P75
i-C12P9
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model
01920197 i-C12P9
01920195 i-C12P75
01920196 i-C12P20
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01920197 Supplier OHAUS Supplier No. 30820355
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Introducing the Courier 1000 Series Shipping Scales, a versatile line of lightweight, durable weighing equipment able to handle everything from general shipping applications for businesses to light weighing jobs for veterinary offices.
Its ultra-compact ABS plastic design combines light weight and durability.
  • Courier scales are exceptionally easy to transport thanks to their integrated handles.
  • Large 0.8 in./20 mm high display works with Dynamic Weighing mode to store data and ensure it is easily accessible when weighing packages and is also ideal for weighing small animals in veterinary use.
  • Battery powered (not included) for use in the office or shop.
  • AC adapter power included.
Specifications
Capacity 9 kg
Display Type LCD
Readability 0.005 kg
Model i-C12P9
Type Scale
Legal for Trade Class III