missing translation for 'onlineSavingsMsg'
Learn More

Coumarin, 99+%

CAS: 91-64-5 | C9H6O2 | 146.15 g/mol

Supplier:  Thermo Scientific Chemicals 110530010

Quantity 1 kg
Packaging Plastic bottle

 Afficher plus de versions de ce produit

Numéro de catalogue. AC110530010


Il en reste null

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Spécifications

Identifiants chimiques

91-64-5
146.15
ZYGHJZDHTFUPRJ-UHFFFAOYSA-N
323
chromen-2-one
C9H6O2
MFCD00006850
2H-1-Benzopyran-2-one
CHEBI:28794
O=C1OC2=CC=CC=C2C=C1

Spécifications

Coumarin
99+%
99+%
MFCD00006850
15, 2550
ZYGHJZDHTFUPRJ-UHFFFAOYSA-N
Authentic
146.15
CHEBI:28794
≥99.0%
White
Crystalline Powder or Crystals
68°C to 71°C
91-64-5
C9H6O2
17, 328
2H-1-Benzopyran-2-one
O=C1OC2=CC=CC=C2C=C1
chromen-2-one
323
146.15
Plastic bottle
1 kg
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
Sécurité et manipulation

Sécurité et manipulation

GHS H Statement:
Causes skin irritation.
Harmful if inhaled.
Harmful in contact with skin.
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Suspected of causing cancer.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 202-086-7

RTECSNumber : GN4200000

TSCA : TSCA

FDS
Documents

Documents

Promotions

Promotions