missing translation for 'onlineSavingsMsg'
Learn More

Coumarin 6 UV-VIS Internal Standard in Methanol, 32 ppm, SPEX CertiPrep™

Catalog No. UV459M
Change view
Click to view available options
Quantity:
3.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
UV459M 3.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. UV459M Supplier SPEX CertiPrep Group LLC Supplier No. UV459M
Only null left

UV-VIS internal standards allow the user to accurately measure the concentration of their samples by directly comparing the absorbance of their compounds against that of the internal standard.

SPEX Certiprep™ offers eight different standards that cover a wide range of wavelengths (270 to 627 nm). After the user takes a UV-VIS spectrum of their sample, they can find a region in the spectrum that doesn’t absorb, and then select the corresponding internal standard, and rerun the spectrum.

Specifications

Chemical Name or Material Coumarin 6
Packaging Ampoule
CAS 38215-36-0
Molecular Formula C20H18N2O2S
Concentration 32 ppm
Technique UV/Vis, Spectrometry, LC/MS, HPLC
Color Colorless to Light Yellow
For Use With (Equipment) UV/Vis Spectrophotometer
Synonym UV-459 nm internal standard
Solubility Soluble in methanol
Solution Type Standard Solution
Type UV Internal Standard
Molecular Weight (g/mol) 350.4
Quantity 3.5 mL
Show More Show Less