missing translation for 'onlineSavingsMsg'
Learn More

Coumarin-3-carboxylic acid, 98%

Catalog No. AAL0713322
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0713322 100 g
AAL0713314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0713322 Supplier Thermo Scientific Chemicals Supplier No. L0713322
Get Up to $1,800 in Chemicals with Purchase
View all promotions

missing translation for 'validMainframeMsg'

CAS: 531-81-7 | C10H6O4 | 190.154 g/mol

Coumarin-3-carboxylic acid is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Coumarin-3-carboxylic acid is used as pharmaceutical intermediate.

Solubility
Soluble in water (13 g/L at 37°C) and alcohol.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 531-81-7
Molecular Formula C10H6O4
Molecular Weight (g/mol) 190.154
MDL Number MFCD00006852
InChI Key ACMLKANOGIVEPB-UHFFFAOYSA-N
Synonym coumarin-3-carboxylic acid, 2-oxo-2h-chromene-3-carboxylic acid, 3-carboxycoumarin, 2-oxobenzopyran-3-carboxylic acid, g 1 rodenticide, 2h-1-benzopyran-3-carboxylic acid, 2-oxo, 2-oxo-2h-1-benzopyran-3-carboxylic acid, unii-v85uov8788, g 1 the rodenticide van, g 1 van
PubChem CID 10752
IUPAC Name 2-oxochromene-3-carboxylic acid
SMILES C1=CC=C2C(=C1)C=C(C(=O)O2)C(=O)O

Specifications

Melting Point 190°C to 193°C
Quantity 100 g
UN Number UN2811
Beilstein 154276
Merck Index 14,2563
Solubility Information Soluble in water (13g/L at 37°C) and alcohol.
Formula Weight 190.15
Percent Purity 98%
Chemical Name or Material Coumarin-3-carboxylic acid
Hazard Category H301
Hazard Statement GHS H Statement
H301
Toxic if swallowed.
Precautionary Statement P264b-P270-P301+P310-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 208-518-0
RTECSNumber DJ2490000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only