missing translation for 'onlineSavingsMsg'
Learn More

Coulter™ Micro Particle Size Standard (Based on polystyrene monodisperse size: 200 m), MilliporeSigma™ Supelco™

Catalog No. 111003742
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111003742 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111003742 Supplier Merck Emd Millipore Supplier No. 070915MLF

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Product Line Coulter
Flash Point 210°C
Purity Grade Notes For every lot exact values of particle size and standard deviation are determined with an accuracy of 0.001 mum. The size determination of these particles follows procedures described by National Institute of Standards and Technology (NIST, USA) respectively by the Community Bureau of Reference (BCR), esp.:. Transmission electron microscopy (TEM). Scanning electron microscope. Light microscopy . Coulter(TM) counter with MDF-systemCalibration of equipment is carried out with NIST-respectively BCR-particle size standards.
Physical Form Aqueous Suspension
Concentration 2% (solids)
MDL Number MFCD00243243
Synonym Size standards
Type Size Standard
Quantity 5 mL

Coulter is a trademark of Beckman Coulter, Inc.