missing translation for 'onlineSavingsMsg'
Learn More

Corning™ 96-Well, Cell Culture-Treated, Flat-Bottom Microplate

Color:White; Sterility:Sterile; Lid:With Lid

Supplier:  Corning™ C3917

Numéro de catalogue. 07200628


Il en reste null

Description

Description

 

  • Minimal well-to-well crosstalk
  • White microplates enhance luminecent signals and have low background luminescence
  • Recommended working volumes of 75 to 200μL
  • Available untreated for solution-based assays, with high binding for immunoassay applications, or tissue culture treated for cell growth
  • Medical-grade polystyrene
  • Well volume: 320μL to 360μL
  • Working volume: 75μL to 200μL
  • Choice of flat, round or V-shaped bottom
  • Treated surface options are not coated, require no special handling and are stable at room temperature
  • Sterile microplates are sterilized by gamma radiation and certified nonpyrogenic
  • Ideal for serial dilutions, hemagglutination or precipitation assays, and general storage applications
  • Plate dimensions meet proposed industry standards

 

Spécifications

Spécifications

Standard
8 x 12
White
Polystyrene
75 to 200 μL
Cell Culture
Attachment and growth of anchorage-dependent cells
With Lid
20/Pk., 100/Cs.
Assay Plate
96
Sterile
Flat Bottom (F-well)
100/Cs.
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
FDS
Documents

Documents

Promotions

Promotions