missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Cortisone, 98%

Numéro de catalogue. p-7077842
Modifier l'affichage
Click to view available options
Quantité:
250mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité
AC460522500 250mg
AC460520010 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. AC460522500 Fournisseur Thermo Scientific™ Code fournisseur 460522500

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Identifiants chimiques

CAS 53-06-5
Formule moléculaire C21H28O5
Poids moléculaire (g/mol) 360.45
Numéro MDL MFCD00003610
Clé InChI MFYSYFVPBJMHGN-ZPOLXVRWSA-N
Synonyme cortisone, andreson, cortisal, cortisate, cortistal, cortivite, cortogen, cortone, kendall's compound e, adrenalex
PubChem CID 222786
ChEBI CHEBI:16962
Nom de l’IUPAC (8S,9S,10R,13S,14S,17R)-17-hydroxy-17-(2-hydroxyacetyl)-10,13-dimethyl-1,2,6,7,8,9,12,14,15,16-decahydrocyclopenta[a]phenanthrene-3,11-dione
SOURIRES C[C@]12CC(=O)[C@H]3[C@@H](CCC4=CC(=O)CC[C@]34C)[C@@H]1CC[C@]2(O)C(=O)CO

Spécifications

Nom ou substance chimique Cortisone
Perte à la dessication 2% max.
Spectre infrarouge Authentic
Fourchette de pourcentages d’essai 97.5% min. (HPLC)
Quantité 250mg
Poids de formule 360.44
Pourcentage de pureté 98%
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Suspected of damaging the unborn child.
Precautionary Statement GHS P Statement
Obtain special instructions before use.
Use personal protective equipment as required.
IF exposed or concerned: Get medical advice/attention.
EINECSNumber 200-162-4

RUO – Research Use Only