missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Synthemax™ II-SC Substrate
GREENER_CHOICE

Catalog No. 07201611
Change view
Click to view available options
Quantity:
10 mg (vial)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
07201611 10 mg (vial)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07201611 Supplier Corning™ Supplier No. C3535
Get Up to $2,500 in Products with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Self-coating synthetic surface for stem cells

Easy-to-use, self-coatingmaterial for creating a unique synthetic surface that mimics the natural cell environment. Surface supportsthe adhesion and expansion of stem cells in their undifferentiated state and directed differentiation into specialized cell types. In addition, it offers a synthetic, xeno-free alternative to biological coatings and/or feeder cell layers used in hES and iPS cell culture.

Allows for the scalable multi-passage expansion of pluripotent stem cells in defined media with:

  • Stable doubling time
  • Phenotypicmarker expression (Oct4, SSEA4)
  • Retention of pluripotency
  • Retention of normal karyotype
Corning Synthemax II-SC Substrate is:
  • Easy-to-use
  • Able to coat plastic and glass surfaces
  • Lot-traceable
  • Gamma sterilized
  • Quality tested for lot-to-lot consistency

Specifications

For Use With (Application) Creating a unique synthetic surface that mimics the natural cell environment
Quantity 10 mg (vial)
Substrate Type Synthemax™ II-SC Substrate
Includes 1 mg/mL Synthemax II SC stock solution, sterile water, 0.025 mg/mL final volume of Corning™ Synthemax II SC working solution
Storage Requirements -20°C
Substrate Quantity 10 mg