missing translation for 'onlineSavingsMsg'
Learn More

Corning™ SOC Medium

Catalog No. MT46003CR
Change view
Click to view available options
Quantity:
10 x 10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MT46003CR 10 x 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MT46003CR Supplier Corning™ Supplier No. 46003CR

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Maximizes transformation efficiency of E. coli competent cells

Store at Room Temperature.

Specifications

For Use With (Application) Use to maximize transformation efficiency of competent E. coli cells
Includes 2% Tryptone, 0.5% Yeast Extract, 0.4% Glucose, 10 mm NaCl, 2.5 mm KCl, 5 mm mgCl2, 5 mm mgSO4
Quantity 10 x 10 mL