missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ Reusable Plastic Reagent Bottles with GL-45 PP Screw Cap
Translucent polypropylene; with polypropylene screw cap.
$236.94 - $435.12
Specifications
| Color | Translucent |
|---|---|
| Material | Polypropylene |
| No. per Pack | 1/Pk. |
| Sterility | Non-sterile |
| Shape | Cylindrical |
| Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity & Availability | |||||
|
07202106
|
Corning™
1500P250 |
250 mL |
Case of 20 Each for $302.14
|
|
|||||
|
07202107
|
Corning™
1500P500 |
500 mL |
Case of 10 Each for $236.94
|
|
|||||
|
07202108
|
Corning™
1500P1L |
1 L |
Case of 10 Each for $435.12
|
|
|||||
|
07202109
|
Corning™
1500P2L |
2 L |
Case of 6 Each for $342.20
|
|
|||||
Description
These bottles are manufactured from translucent polypropylene. They are wide-mouth and supplied with PP screw top caps.
Specifications
| Translucent | |
| 1/Pk. | |
| Cylindrical |
| Polypropylene | |
| Non-sterile |