missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Reusable Plastic Reagent Bottles with GL-45 PP Screw Cap

Catalog No. 07202109
Change view
Click to view available options
Capacity (Metric):
250 mL
500 mL
1 L
2 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
07202109 2 L
07202106 250 mL
07202107 500 mL
07202108 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 07202109 Supplier Corning™ Supplier No. 1500P2L

Translucent polypropylene; with polypropylene screw cap.

These bottles are manufactured from translucent polypropylene. They are wide-mouth and supplied with PP screw top caps.

Specifications

Capacity (Metric) 2 L
Color Translucent
Material Polypropylene
No. per Case 6/Cs.
No. per Pack 1/Pk.
Sterility Non-sterile
Shape Cylindrical
Height (Metric) 24.3 cm
Diameter (Metric) 13.1 cm