missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Reusable Plastic Reagent Bottles with GL-45 PP Screw Cap

Translucent polypropylene; with polypropylene screw cap.

Supplier:  Corning™ 1500P1L

 View more versions of this product

Catalog No. 07202108


Only null left

Description

Description

These bottles are manufactured from translucent polypropylene. They are wide-mouth and supplied with PP screw top caps.

Specifications

Chemical Identifiers

Translucent
1/Pk.
Cylindrical
Polypropylene
Non-sterile

Specifications

1 L
Polypropylene
1/Pk.
Cylindrical
10.8 cm
Translucent
10/Cs.
Non-sterile
20.4 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions