Learn More
Corning™ PureCoat™ Carboxyl Multiwell Plates
Numéro de catalogue. 08772176
Change view
Click to view available options
Well Count:
6
Numéro de catalogue. Well Count
08772176 6
1 options
Numéro de catalogue. 08772176 Fournisseur Corning™ Code fournisseur 356773
Il en reste null
Corning PureCoat Amine (positively charged) and Carboxyl (negatively charged) surfaces provide improved cell attachment, faster cell proliferation, and enhanced recovery post-thaw over standard TC surfaces. These surfaces function with a broad range of primary, transfected, transformed, and fastidious cell types, and have demonstrated utility in serum-reduced or serum-free conditions.
Corning PureCoat Amine (positively charged) and Carboxyl (negatively charged) surfaces provide improved cell attachment, faster cell proliferation, and enhanced recovery post-thaw over standard TC surfaces. These surfaces function with a broad range of primary, transfected, transformed, and fastidious cell types, and have demonstrated utility in serum-reduced or serum-free conditions.
Spécifications
Array 2 x 3
Certifications/Compliance ANSI, SBS
Bottom Shape Flat
Material Polystyrene
Shape Rectangular
Sterility Sterile
Type Multi-well Cell Culture Plate
Well Count 6
For Use With (Application) Culture of primary cells, transfected, transformed and fastidious cell lines, culture in standard, serum-free or serum-reduced conditions, freeze-thaw recovery of cryopreserved primary cells and cell lines, including division arrested
No. per Case 50/Cs.
No. per Pack 5/Pk.
Quantity 50/Cs.
Afficher plus de résultats Afficher moins de résultats