Learn More
Corning™ LSE™ Platform Rocker
Catalog No. 07202202
Change view
Click to view available options
Model:
Double level
Single level
Plug Type:
EU Plug
US Plug
Voltage:
115 V
230 V
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model Plug Type Voltage
07202202 Double level US Plug 115 V
07202201 Single level EU Plug 230 V
07202203 Double level EU Plug 230 V
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 07202202 Supplier Corning™ Supplier No. 6705
Get Up to $2,500 in Products with Purchase
View all promotions
Edge
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Corning LSE Platform Rockers are ideal for Western Blotting, Gel staining and general mixing applications. Comes in single and double (30 x 30 cm) platform models to provide extra capacity.
The Corning™ LSE™ Platform Rocker is a compact yet high capacity rocker that is ideal for western blotting and general mixing applications. The adjustable rocking speed and fixed 7 degree tilt makes it optimal for staining fragile gels, as well as the more intensive action necessary for washing blots.
The speed of the Corning Platform Rocker can be adjusted from 2 to 60 rpm and the timer can be set for up to 2 hours or for continuous operation. The unit is safe to use in temperature controlled environments up to 70°C and is available in single or high capacity double platform configurations. Both of these rugged models come with a 5-year warranty.
Features and Benefits
  • Low profile and compact size
  • 11-pound load capacity
  • Non-slip rubber pads to prevent sample containers from moving during operation
  • Double platform model available for increased capacity
See resources below for sell sheet with specifications.
Specifications
Voltage 115 V
Product Type Platform Rocker
Plug Type US Plug
Model Double level