missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Low-Volume 384-Well Polystyrene Plates
GREENER_CHOICE

Catalog No. 0976184
Change view
Click to view available options
Surface Treatment:
Non-binding
Non-treated
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Surface Treatment
0976184 Non-binding
07202625 Non-treated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 0976184 Supplier Corning™ Supplier No. 3544

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

High-base design is compatible with automation and bar coding

  • Validated for the FLIPR™ TETRA system—outer wells aligned for proper viewing with the FLIPR system
  • 2.8mm distance between well bottoms and plate bottom
  • Round wells with flat bottoms
  • Working volume: 5–45μL

  • Untreated surface binds biomolecules through passive interactions; Nonbinding surface (NBS™) creates a nonionic hydrophilic surface that minimizes molecular interactions; Tissue culture-treated surface provides optimal cell attachment

Specifications

Format Standard
Lid Without Lid
Array 16 x 24
Color Black
Material Polystyrene
No. of Wells 384
Volume (Metric) Working 0.005 to 0.04 mL
Sterility Non-sterile
Surface Treatment Non-binding
Well Shape Flat Bottom (F-well)
For Use With (Application) For miniaturized 384-well fluorescent homogeneous assays
Show More Show Less