missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ Dispase

Description
Corning extracellular matrices (ECMs) enable researchers to mimic in vivo environments for 2D and 3D cell culture applications.
Spécifications
Spécifications
| For Use With (Application) | Recovering cells cultured on BD Matrigel™ basement membrane matrix |
| Content And Storage | -20°C |
| Source | Bacillus polymyxa Metallo Protease |
| Quantity | 100 mL |
| Product Type | Dispase |
| Molecular Weight (g/mol) | 36 |