missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Collagen IV, Mouse
GREENER_CHOICE

Numéro de catalogue. CB40233
Modifier l'affichage
Click to view available options
Quantité:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité
CB40233 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. CB40233 Fournisseur Corning™ Code fournisseur 354233

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Used as either a thin layer on tissue-culture surfaces to enhance cell attachment and proliferation, or as a gel to promote expression of cell-specific morphology and function

Corning extracellular matrices (ECMs) enable researchers to mimic in vivo environments for 2D and 3D cell culture applications.

Corning offers a broad range of collagen types that are derived from multiple species. These collagens have been used to culture a variety of cell types to promote adhesion, growth, and/or differentiation.

Spécifications

Concentration 1 to 5 μg/cm2
À utiliser avec (application) Cell Attachment and Proliferation
Formule Frozen in 0.05 M Hydrochloric Acid (HCL)
Quantité 1 mg
Type de produit Collagen, Mouse Type IV
Contenu et stockage -70°C