missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Collagen I, Rat
GREENER_CHOICE

Numéro de catalogue. CB40236
Modifier l'affichage
Click to view available options
Quantité:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité
CB40236 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. CB40236 Fournisseur Corning™ Code fournisseur 354236

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Enhances cell attachment and proliferation or promotes expression of cell-specific morphology and function. Corning™ Collagen I, Rat is effective as a gel or a thin coating at a recommended concentration of 5 to 10μg/cm2 of growth surface depending on cell type.

Corning extracellular matrices (ECMs) enable researchers to mimic in vivo environments for 2D and 3D cell culture applications.

Corning offers a broad range of collagen types that are derived from multiple species. These collagens have been used to culture a variety of cell types to promote adhesion, growth, and/or differentiation.

Spécifications

Concentration 3 to 4 mg/mL
À utiliser avec (application) Cell Attachment and Proliferation, Cell-specific Morphology and Function
Formule Liquid in 0.02N acetic acid
Quantité 100 mg
Sources Rat Tail Tendon
Type de produit Collagen, Rat Tail Type I
Contenu et stockage 2°C to 8°C
Grade de qualité ou de pureté >90% by SDS-PAGE