missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Collagen I, High Concentration, Rat Tail
GREENER_CHOICE

Catalog No. CB354249
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CB354249 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CB354249 Supplier Corning™ Supplier No. 354249

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Sourced from rat tail tendon

Corning extracellular matrices (ECMs) enable researchers to mimic in vivo environments for 2D and 3D cell culture applications.

Corning offers a broad range of collagen types that are derived from multiple species. These collagens have been used to culture a variety of cell types to promote adhesion, growth, and/or differentiation.

Specifications

Concentration 8 to 11 mg/mL
For Use With (Application) Cell Attachment and Proliferation, 3D Gel
Formulation Liquid in 0.02N acetic acid
Quantity 100 mg
Source Rat Tail Tendon
Product Type Collagen, Rat Tail Type I HC
Content And Storage 2°C to 8°C
Purity or Quality Grade >95% by SDS-PAGE

For Research Use Only (RUO).