missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Gentamicin Sulfate

Numéro de catalogue. MT30005CR
Change view
Click to view available options
Quantity:
1 x 100 mg
1 x 1 g
10 x 10 mL
Form:
Liquid
Powder
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity Form
MT30005CR 10 x 10 mL Liquid
MT61098RA 1 x 100 mg Powder
MT61098RF 1 x 1 g Powder
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Numéro de catalogue. MT30005CR Fournisseur Corning™ Code fournisseur 30005CR
Il en reste null

Packaged in plastic bottles

Spécifications

Content And Storage 15° to 30°C
Concentration 50 mg/mL
Form Liquid
Product Type Antibiotic
Sterility Sterile
Format Plastic Bottle
Quantity 10 x 10 mL