missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ BioCoat™ Collagen IV Multiwell Plates

Description
- Type IV Collagen is a ubiquitous component in basement membranes and provides the major structural support for this matrix. When the Collagen IV meshwork is assembled, it provides a scaffold for the assembly of other basement membrane components through interactions with laminin, entactin/nidogen, and heparan sulfate proteoglycan. Collagen IV is useful as a substrate for growth of epithelial, endothelial, muscle, and nerve cells. Collagen plays a role in the regulation of cell growth, differentiation and adhesion, as well as tissue formation.
Applications include:
- Promotion of cell attachment and spreading
- Cell differentiation and neurite outgrowth
- Increased proliferation of PC12 cells
- Studies of effects of collagen IV on cell behavior
- Cell adhesion assays
Source
- Engelbreth-Holm-Swarm (EHS) lathrytic mouse tumor
Quality Control
- Tested for ability to initiate differentiation (neurite outgrowth) of NG-108 rat glioma/mouse neuroblastoma cells
- Tested and found negative for bacteria and fungi
- Collagen IV purity >90% by SDS-PAGE
Storage
- 2°C to 8°C. Do not freeze.
Spécifications
Spécifications
| Array | 2 x 3 |
| Closure Color | Clear |
| Closure Included | Yes |
| Closure Material | Polystyrene |
| Bottom Shape | Flat |
| Color | Clear |
| Material | Polystyrene |
| Shape | Rectangular |
| Sterility | Non-sterile |
| Surface Treatment | ECM/Attachment Factors |
| Afficher plus de résultats |