missing translation for 'onlineSavingsMsg'
Learn More

Corning™ 96-Well Nonbinding Surface (NBS™) Microplates
GREENER_CHOICE

Treated with a Corning proprietary treatment technology to create a nonionic hydrophilic surface (polyethylene oxide-like) that minimizes molecular interactions. Corning™ 96-Well Nonbinding Surface (NBS™) Microplates are ideal for reducing proteing and nucleic acid binding at low concentrations, and increasing assay signal to noise.

$2,242.51 - $3,283.48

Spécifications

Format Standard
Lid Without Lid
Array 8 x 12
For Use With (Equipment) Plate readers, liquid handlers and robotic systems
Material Polystyrene
voir les spécifications

Produits 4
Numéro de catalogue Numéro du manufacturier. Color Well Shape Prix Quantity & Availability  
Numéro de catalogue Numéro du manufacturier. Color Well Shape Prix Quantity & Availability  
07200731
GREENER_CHOICE
Afficher les documents Détails de la promotion
Corning™
3600
White Flat Bottom (F-well)
caisse de 100 chaque for $3,200.77
Il en reste null
 
07200777
GREENER_CHOICE
Accessoires Afficher les documents Détails de la promotion
Corning™
3641
Clear Flat Bottom (F-well)
caisse de 100 chaque for $2,242.51
Il en reste null
 
07200778
GREENER_CHOICE
Afficher les documents Détails de la promotion
Corning™
C3650
Black Flat Bottom (F-well)
caisse de 100 chaque for $3,283.48
Il en reste null
 
07200791
GREENER_CHOICE
Afficher les documents Détails de la promotion
Corning™
C3605
White Round
caisse de 100 chaque for $3,100.91
Il en reste null
 
Description

Description

  • Designed to reduce well-to-well cross-talk
  • White microplates enhance luminescent signals and have low background luminescence.
  • Black microplates have low background fluorescence and minimize light scattering.
  • Spécifications

    Spécifications

    Standard
    8 x 12
    Polystyrene
    75 to 200 μL
    Non-binding
    100/Cs.
    100/Cs.
    4° to 37°C
    Without Lid
    Plate readers, liquid handlers and robotic systems
    96
    Non-sterile
    For homogenous assays such as scintillation proximity assays (SPA) and fluorescent assays
    25/Pk.
    39°F to 99°F
    Videos
    FDS
    Documents

    Documents