missing translation for 'onlineSavingsMsg'
Learn More

Corning™ 96-Well Nonbinding Surface (NBS™) Microplates
GREENER_CHOICE

Catalog No. 07200777
Change view
Click to view available options
Color:
Black
Clear
White
Well Shape:
Flat Bottom (F-well)
Round
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Well Shape
07200777 Clear Flat Bottom (F-well)
07200731 White Flat Bottom (F-well)
07200778 Black Flat Bottom (F-well)
07200791 White Round
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 07200777 Supplier Corning™ Supplier No. 3641

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Treated with a Corning proprietary treatment technology to create a nonionic hydrophilic surface (polyethylene oxide-like) that minimizes molecular interactions. Corning™ 96-Well Nonbinding Surface (NBS™) Microplates are ideal for reducing proteing and nucleic acid binding at low concentrations, and increasing assay signal to noise.

  • Designed to reduce well-to-well cross-talk
  • White microplates enhance luminescent signals and have low background luminescence.
  • Black microplates have low background fluorescence and minimize light scattering.
  • Specifications

    Format Standard
    Lid Without Lid
    Array 8 x 12
    Packaging 4 inner packs of 25
    Binding Type NBS
    Color Clear
    For Use With (Equipment) Plate readers, liquid handlers and robotic systems
    Material Polystyrene
    No. of Wells 96
    Volume (Metric) Working 75 to 200 μL
    Sterility Non-sterile
    Surface Treatment Non-binding
    Well Shape Flat Bottom (F-well)
    For Use With (Application) For homogenous assays such as scintillation proximity assays (SPA) and fluorescent assays
    No. per Case 100/Cs.
    No. per Pack 25/Pk.
    Quantity 100/Cs.
    Temperature Range (English) 39°F to 99°F
    Temperature Range (Metric) 4° to 37°C
    Well Color Clear
    Show More Show Less