missing translation for 'onlineSavingsMsg'
Learn More

Corning™ 96-Well Nonbinding Surface (NBS™) Microplates
GREENER_CHOICE

Catalog No. 07200791
Change view
Click to view available options
Color:
Black
Clear
White
Well Shape:
Flat Bottom (F-well)
Round
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Well Shape
07200791 White Round
07200731 White Flat Bottom (F-well)
07200777 Clear Flat Bottom (F-well)
07200778 Black Flat Bottom (F-well)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 07200791 Supplier Corning™ Supplier No. C3605
Get Up to $2,500 in Products with Purchase
View all promotions

Treated with a Corning proprietary treatment technology to create a nonionic hydrophilic surface (polyethylene oxide-like) that minimizes molecular interactions. Corning™ 96-Well Nonbinding Surface (NBS™) Microplates are ideal for reducing proteing and nucleic acid binding at low concentrations, and increasing assay signal to noise.

  • Designed to reduce well-to-well cross-talk
  • White microplates enhance luminescent signals and have low background luminescence.
  • Black microplates have low background fluorescence and minimize light scattering.
  • Specifications

    Format Standard
    Lid Without Lid
    Array 8 x 12
    Color White
    For Use With (Equipment) Plate readers, liquid handlers and robotic systems
    Material Polystyrene
    No. of Wells 96
    Volume (Metric) Working 75 to 200 μL
    Sterility Non-sterile
    Surface Treatment Non-binding
    Well Shape Round
    For Use With (Application) For homogenous assays such as scintillation proximity assays (SPA) and fluorescent assays
    No. per Case 100/Cs.
    No. per Pack 25/Pk.
    Quantity 100/Cs.
    Temperature Range (English) 39°F to 99°F
    Temperature Range (Metric) 4° to 37°C
    Well Color White
    Show More Show Less