missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Universal CellSTACK™ Caps

Catalog No. 07201521
Change view
Click to view available options
Closure Size:
33 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Closure Size
07201521 33 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07201521 Supplier Corning™ Supplier No. 3334
Get Up to $2,500 in Products with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For filling, venting, and manipulation

  • 33 mm threaded cap
  • Solid cap is used in closed systems or applications where venting is not necessary
  • Sterilized by gamma irradiation

Specifications

Closure Color Orange
Closure Material HDPE
Color Orange
Material HDPE
Includes Male MPC Polycarbonate, 0.25 in. I.D. coupling and female MPC Polycarbonate end cap
No. per Case 4/Cs.
No. per Pack 1/Pk.
Product Type Cap, Filling
Disposable Single-use
Sterility Sterile
Closure Type Screw Cap
Closure Size 33 mm
Show More Show Less