missing translation for 'onlineSavingsMsg'
Learn More

Cordycepin from Cordyceps militaris 98.0+%, TCI America™

Catalog No. C268925MG
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C268925MG 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C268925MG Supplier TCI America Supplier No. C268925MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 73-03-0
Molecular Formula C10H13N5O3
Molecular Weight (g/mol) 251.25
MDL Number MFCD00037998
InChI Key OFEZSBMBBKLLBJ-PNFQIUPPNA-N
Synonym cordycepin, 3'-deoxyadenosine, cordycepine, 9-cordyceposidoadenine, adenosine, 3'-deoxy, 9-beta-d-3'-deoxyribofuranosyl adenine, unii-gz8vf4m2j8, 9h-purine, 6-amino-9-3-deoxy-beta-d-ribofuranosyl, ccris 3233, gz8vf4m2j8
PubChem CID 6303
ChEBI CHEBI:29014
IUPAC Name (2R,3R,5S)-2-(6-amino-9H-purin-9-yl)-5-(hydroxymethyl)oxolan-3-ol
SMILES NC1=C2N=CN([C@@H]3O[C@H](CO)C[C@H]3O)C2=NC=N1

Specifications

Melting Point 226°C
Color White-Yellow
Quantity 25 mg
Formula Weight 251.25
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Cordycepin from Cordyceps militaris
RTECSNumber AU7358610
TSCA No
Recommended Storage Refrigerator