missing translation for 'onlineSavingsMsg'
Learn More

Copper Tetrammine, Ammoniacal Solution, Ricca Chemical

Catalog No. R2370000250
Change view
Click to view available options
Quantity:
250 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R2370000250 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R2370000250 Supplier Ricca Chemical Company Supplier No. R2370000250A
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

EP Reagent 1022600

Chemical Identifiers

CAS 1336-21-6 , 7732-18-5 , 7758-99-8 , 1310-73-2
Molecular Formula H5NO
Molecular Weight (g/mol) 35.05
MDL Number MFCD00066650
InChI Key VHUUQVKOLVNVRT-UHFFFAOYSA-N
Synonym ammonium hydroxide, ammonia water, ammonia aqueous, aquammonia, aqua ammonia, ammonia, aqua, household ammonia, ammonium hydroxide solution, ammonia, monohydrate, ammonium hydroxide nh4 oh
PubChem CID 14923
ChEBI CHEBI:18219
IUPAC Name amine hydrate
SMILES N.O

Specifications

Color Blue
CAS Min % 54.0, 30.67, 8.66, 4.49
CAS Max % 56.0, 31.93, 9.01, 4.67
Grade Laboratory
Packaging Natural Poly Bottle
pH Alkaline
Quantity 250 mL
Physical Form Liquid
Chemical Name or Material Copper Tetrammine, Ammoniacal Solution