missing translation for 'onlineSavingsMsg'
Learn More

Copper Sulfate, 1.053 Specific Gravity at 25°C (77°F), Ricca Chemical

Catalog No. 233032
Change view
Click to view available options
Package: Amber Glass; Size: 1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
233032 Package: Amber Glass; Size: 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 233032 Supplier Ricca Chemical Company Supplier No. 233032

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For Hemoglobin Estimation in Blood Donor Screening of Men and Women

Specifications

CAS 7758-99-8, 7732-18-5
CAS Min % 90.0, 7.52
CAS Max % 94.0, 7.83
Molecular Formula CuH10O9S
MDL Number MFCD00149681
Synonym copper ii sulfate pentahydrate, copper sulfate pentahydrate, cupric sulfate pentahydrate, blue vitriol, calcanthite, bluestone, copper 2+ sulfate pentahydrate, triangle, vencedor, blue copperas
InChI Key JZCCFEFSEZPSOG-UHFFFAOYSA-L
SMILES O.O.O.O.O.[Cu++].[O-]S([O-])(=O)=O
IUPAC Name copper(2+) pentahydrate sulfate
Molecular Weight (g/mol) 249.68
PubChem CID 24463
ChEBI CHEBI:31440
Packaging Amber Glass Bottle
Quantity 1 L
Chemical Name or Material Copper Sulfate Solution, Specific Gravity 1.053
Show More Show Less