missing translation for 'onlineSavingsMsg'
Learn More

Copper Sulfate Solution, for Aluminum Can Coating Test, Ricca Chemical

Catalog No. 23261
Change view
Click to view available options
Quantity:
4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23261 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 23261 Supplier Ricca Chemical Company Supplier No. RI23261

For Aluminum Can Coating Test

Chemical Identifiers

CAS 7758-99-8 , 7732-18-5 , 7647-01-0
Molecular Formula CuH10O9S
Molecular Weight (g/mol) 249.68
MDL Number MFCD00149681
InChI Key JZCCFEFSEZPSOG-UHFFFAOYSA-L
Synonym copper ii sulfate pentahydrate, copper sulfate pentahydrate, cupric sulfate pentahydrate, blue vitriol, calcanthite, bluestone, copper 2+ sulfate pentahydrate, triangle, vencedor, blue copperas
PubChem CID 24463
ChEBI CHEBI:31440
IUPAC Name copper(2+) pentahydrate sulfate
SMILES O.O.O.O.O.[Cu++].[O-]S([O-])(=O)=O

Specifications

Color Blue
CAS Min % 16.78, 73.0, 8.65
CAS Max % 17.46, 76.0, 9.0
Grade Laboratory
Packaging Amber Glass Bottle
pH 1.98 to 2.16
Quantity 4 L
Physical Form Liquid
Chemical Name or Material Copper Sulfate Solution