missing translation for 'onlineSavingsMsg'
Learn More

Copper Sulfate, 20% (w/v), Ricca Chemical

$117.53 - $438.23

Chemical Identifiers

CAS 7758-99-8
Molecular Formula CuH10O9S
Molecular Weight (g/mol) 249.68
MDL Number MFCD00149681
InChI Key JZCCFEFSEZPSOG-UHFFFAOYSA-L
Synonym copper ii sulfate pentahydrate, copper sulfate pentahydrate, cupric sulfate pentahydrate, blue vitriol, calcanthite, bluestone, copper 2+ sulfate pentahydrate, triangle, vencedor, blue copperas
PubChem CID 24463
ChEBI CHEBI:31440
IUPAC Name copper(2+) pentahydrate sulfate
SMILES O.O.O.O.O.[Cu++].[O-]S([O-])(=O)=O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
2325-16
SDS View Documents
Ricca Chemical Company
232516
500 mL Natural Poly Bottle
Each for $117.53
Only null left
 
2325-32
View Documents
Ricca Chemical Company
232532
1 L Natural Poly Bottle
Each for $164.20
Only null left
 
2325-1
View Documents
Ricca Chemical Company
23251
4 L Natural Poly Bottle
Each for $438.23
Only null left
 
Description

Description

Specifications

Chemical Identifiers

7758-99-8
249.68
JZCCFEFSEZPSOG-UHFFFAOYSA-L
24463
copper(2+) pentahydrate sulfate
CuH10O9S
MFCD00149681
copper ii sulfate pentahydrate, copper sulfate pentahydrate, cupric sulfate pentahydrate, blue vitriol, calcanthite, bluestone, copper 2+ sulfate pentahydrate, triangle, vencedor, blue copperas
CHEBI:31440
O.O.O.O.O.[Cu++].[O-]S([O-])(=O)=O

Specifications

Blue
82.0,19.0
CuH10O9S
copper ii sulfate pentahydrate, copper sulfate pentahydrate, cupric sulfate pentahydrate, blue vitriol, calcanthite, bluestone, copper 2+ sulfate pentahydrate, triangle, vencedor, blue copperas
O.O.O.O.O.[Cu++].[O-]S([O-])(=O)=O
249.68
CHEBI:31440
Laboratory
Aqueous Solution
500 mL
20% (w/v)
7758-99-8,7732-18-5
85.0,21.0
MFCD00149681
JZCCFEFSEZPSOG-UHFFFAOYSA-L
copper(2+) pentahydrate sulfate
24463
20%
Natural Poly Bottle
∼4
Liquid
Copper Sulfate
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'