missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper Sulfate, 20% (w/v), Ricca Chemical
Specifications
Specifications
| Color | Blue |
| CAS Min % | 82.0, 19.0 |
| CAS Max % | 85.0, 21.0 |
| Percent Purity | 20% |
| Grade | Laboratory |
| Packaging | Natural Poly Bottle |
| Solvent or Matrix | Aqueous Solution |
| pH | ∼4 |
| Quantity | 500 mL |
| Physical Form | Liquid |
| Show More |