missing translation for 'onlineSavingsMsg'
Learn More

Copper Sulfate, 20% (w/v), Ricca Chemical

Supplier:  Ricca Chemical Company 23251

 View more versions of this product

Catalog No. 2325-1


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Copper Sulfate
7758-99-8,7732-18-5
85.0,21.0
MFCD00149681
JZCCFEFSEZPSOG-UHFFFAOYSA-L
copper(2+) pentahydrate sulfate
24463
20%
Natural Poly Bottle
∼4
Liquid
Blue
82.0,19.0
CuH10O9S
copper ii sulfate pentahydrate, copper sulfate pentahydrate, cupric sulfate pentahydrate, blue vitriol, calcanthite, bluestone, copper 2+ sulfate pentahydrate, triangle, vencedor, blue copperas
O.O.O.O.O.[Cu++].[O-]S([O-])(=O)=O
249.68
CHEBI:31440
Laboratory
Aqueous Solution
4 L
20% (w/v)

Chemical Identifiers

7758-99-8
249.68
JZCCFEFSEZPSOG-UHFFFAOYSA-L
24463
copper(2+) pentahydrate sulfate
CuH10O9S
MFCD00149681
copper ii sulfate pentahydrate, copper sulfate pentahydrate, cupric sulfate pentahydrate, blue vitriol, calcanthite, bluestone, copper 2+ sulfate pentahydrate, triangle, vencedor, blue copperas
CHEBI:31440
O.O.O.O.O.[Cu++].[O-]S([O-])(=O)=O
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions