missing translation for 'onlineSavingsMsg'
Learn More

Copper Sulfate, 10% (w/v), Ricca Chemical

Catalog No. 232016
Change view
Click to view available options
Quantity:
500 mL
4 L
Packaging:
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
232016 500 mL Natural Poly Bottle
23201 4 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 232016 Supplier Ricca Chemical Company Supplier No. 232016

Meets EPA Specifications

Chemical Identifiers

CAS 7758-99-8 , 7732-18-5
Molecular Formula CuH10O9S
Molecular Weight (g/mol) 249.68
MDL Number MFCD00149681
InChI Key JZCCFEFSEZPSOG-UHFFFAOYSA-L
Synonym copper ii sulfate pentahydrate, copper sulfate pentahydrate, cupric sulfate pentahydrate, blue vitriol, calcanthite, bluestone, copper 2+ sulfate pentahydrate, triangle, vencedor, blue copperas
PubChem CID 24463
ChEBI CHEBI:31440
IUPAC Name copper(2+) pentahydrate sulfate
SMILES O.O.O.O.O.[Cu++].[O-]S([O-])(=O)=O

Specifications

Color Blue
CAS Min % 89.0, 9.0
CAS Max % 93.0, 11.0
Percent Purity 10%
Grade Laboratory
Packaging Natural Poly Bottle
Solvent or Matrix Aqueous Solution
pH ∼4
Quantity 500 mL
Physical Form Liquid
Concentration or Composition Notes 10% (w/v)
Chemical Name or Material Copper Sulfate
Show More Show Less