missing translation for 'onlineSavingsMsg'
Learn More

Copper Nickel wire, 0.13mm (0.005in) dia

Catalog No. AA43270H9
Change view
Click to view available options
Quantity:
100 m
500 m
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA43270H9 500 m
AA43270H4 100 m
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA43270H9 Supplier Thermo Scientific Chemicals Supplier No. 043270H9

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Form: Approx. 0.11g/m | Cu:Ni 55:45 wt%

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Color Silver
Chemical Name or Material Copper Nickle
Appearance Silver-colored wire
Quantity 500 m
Odor Odorless
Form Wire
Molecular Formula Cu:Ni 55:45 wt%
MDL Number MFCD00801098

RUO – Research Use Only