missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) sulfate, anhydrous, Reagent Grade

Catalog No. AA3330822
Change view
Click to view available options
Quantity:
100 g
500 g
2 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3330822 100 g
AA3330836 500 g
AA33308A3 2 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA3330822 Supplier Thermo Scientific Chemicals Supplier No. 03330822

CAS: 7758-98-7 | CuO4S | 159.602 g/mol

Copper(II)sulfate is used in electroplating and in mining industries. It is used as a dehydrating agent for forming and manipulating acetal groups. In analytical chemistry, it is used in the Fehling's solution and Benedict's solution to test for reducing sugars. Also, it is used to test protein in Biuret test.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7758-98-7
Molecular Formula CuO4S
Molecular Weight (g/mol) 159.602
MDL Number MFCD00010981
InChI Key ARUVKPQLZAKDPS-UHFFFAOYSA-L
Synonym copper sulfate, copper ii sulfate, cupric sulfate anhydrous, copper sulphate, copper 2+ sulfate, blue stone, copper monosulfate, copper ii sulfate, anhydrous, hylinec, trinagle
PubChem CID 24462
ChEBI CHEBI:23414
IUPAC Name copper;sulfate
SMILES [O-]S(=O)(=O)[O-].[Cu+2]

Specifications

Melting Point >560°C (decomposition)
Color Gray
Physical Form Powder
Quantity 100 g
Solubility Information Soluble in water and methanol. Insoluble in ethanol.
Formula Weight 159.6
Grade Reagent
Odor Odorless
Sensitivity Hygroscopic
Density 3.6
Chemical Name or Material Copper(II) sulfate
Show More Show Less
Hazard Category H302-H315-H319
Precautionary Statement P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P330-P332+P313-P362-P501c
DOTInformation DOT Class: 9; Packaging Group: III
EINECSNumber 231-847-6
RTECSNumber GL8800000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only