missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) oxide, 99+%

Catalog No. AC221281000
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC221281000 100 g Plastic bottle
AC221285000 500 g Plastic bottle
AC221280025 2.5 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC221281000 Supplier Thermo Scientific Chemicals Supplier No. 221281000

CAS: 1317-38-0 | CuO | 79.55 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1317-38-0
Molecular Formula CuO
Molecular Weight (g/mol) 79.55
MDL Number MFCD00010979
InChI Key KKCXRELNMOYFLS-UHFFFAOYSA-N
Synonym cupric oxide, copper ii oxide, copper oxide, copper monoxide, banacobru ol, chrome brown, copper brown, copper monooxide, black copper oxide
PubChem CID 14829
SMILES [O--].[Cu++]

Specifications

Melting Point 1326.0°C
Color Black
Physical Form Powder
Quantity 100 g
Assay Percent Range 99% min. (Complexometry)
Linear Formula CuO
Fieser 04,108; 05,161
Merck Index 15, 2636
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in dil.acids - slowly soluble in nh3, soluble in alkali cyanides and (nh4)2co3 solutions, practically insoluble in alcohol
IUPAC Name oxocopper
Formula Weight 79.54
Percent Purity ≥99%
Packaging Plastic bottle
Chemical Name or Material Copper(II) oxide, p.a.
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Very toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Avoid release to the environment.
Collect spillage.
Dispose of contents/container to an approved waste disposal plant.
EINECSNumber 215-269-1
RTECSNumber GL7900000
TSCA TSCA

RUO – Research Use Only