missing translation for 'onlineSavingsMsg'
Learn More

Copper(II)-ethylenediamine complex, technical, 1M sol. in water, not for viscosity determination

Catalog No. AC460680010
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC460680010 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC460680010 Supplier Thermo Scientific Chemicals Supplier No. 460680010
Only null left

CAS: 14552-35-3 | C4H18CuN4O2 | 217.76 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 14552-35-3 , 7732-18-5
Molecular Formula C4H18CuN4O2
Molecular Weight (g/mol) 217.76

Specifications

Quantity 1 L
Linear Formula Cu(H2NCH2CH2NH2)2(OH)2
Flash Point >110°C
Solubility Information Solubility in water: soluble.
Formula Weight 217.76
Grade Technical
Concentration or Composition (by Analyte or Components) 0.98 to 1.02 M (copper molarity)
Specific Gravity 1.1
Density 1.1000g/mL
Chemical Name or Material Copper(II)-ethylenediamine complex
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
EINECSNumber 238-597-7

RUO – Research Use Only