missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) Dimethyldithiocarbamate 98.0+%, TCI America™

Catalog No. D102225G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D102225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D102225G Supplier TCI America Supplier No. D102225G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 137-29-1
Molecular Formula C6H12CuN2S4
Molecular Weight (g/mol) 303.962
MDL Number MFCD00050845
InChI Key ZOUQIAGHKFLHIA-UHFFFAOYSA-L
Synonym Dimethyldithiocarbamic Acid Copper(II) Salt
PubChem CID 472181
IUPAC Name copper;N,N-dimethylcarbamodithioate
SMILES CN(C)C(=S)[S-].CN(C)C(=S)[S-].[Cu+2]

Specifications

Quantity 25 g
UN Number 3077
Formula Weight 303.96
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Copper(II) Dimethyldithiocarbamate
EINECSNumber (2)-1833
RTECSNumber FA0175000
TSCA Yes