missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) Bis(2-hydroxyethyl)dithiocarbamate 98.0+%, TCI America™

Catalog No. B04915G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B04915G 5 g
B049125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B04915G Supplier TCI America Supplier No. B04915G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 52611-57-1
Molecular Formula C10H20CuN2O4S4
Molecular Weight (g/mol) 424.066
MDL Number MFCD00059149
InChI Key CWZKOKSQKMDNMW-UHFFFAOYSA-L
Synonym Bis(2-hydroxyethyl)dithiocarbamic Acid Copper(II) Salt
PubChem CID 504564
IUPAC Name copper;N,N-bis(2-hydroxyethyl)carbamodithioate
SMILES C(CO)N(CCO)C(=S)[S-].C(CO)N(CCO)C(=S)[S-].[Cu+2]

Specifications

Color Black-Green
Quantity 5 g
Formula Weight 424.07
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Copper(II) Bis(2-hydroxyethyl)dithiocarbamate