missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) 2,3,9,10,16,17,23,24-Octafluorophthalocyanine (purified by sublimation) 98.0+%, TCI America™

Catalog No. C24271G
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C24271G 1 g
C2427100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C24271G Supplier TCI America Supplier No. C24271G

Chemical Identifiers

CAS 148651-60-9
Molecular Formula C32H8CuF8N8
Molecular Weight (g/mol) 720.005
InChI Key SYOZEEBBXMOLOS-UHFFFAOYSA-N
Synonym 2,3,9,10,16,17,23,24-Octafluorophthalocyanine Copper(II), F8CuPc
PubChem CID 53384437
SMILES C1=C2C(=CC(=C1F)F)C3=NC4=NC(=NC5=NC(=NC6=C7C=C(C(=CC7=C([N-]6)N=C2[N-]3)F)F)C8=CC(=C(C=C85)F)F)C9=CC(=C(C=C94)F)F.[Cu+2]

Specifications

Color Blue-Purple
Quantity 1 g
Formula Weight 720.01
Percent Purity ≥98.0% (N,T)
Physical Form Crystalline Powder
Chemical Name or Material Copper(II) 2,3,9,10,16,17,23,24-Octafluorophthalocyanine (purified by sublimation)