missing translation for 'onlineSavingsMsg'
Learn More

Copper ICP-MS Standard, 10,000 ppm Cu in 5% HNO3, Ricca Chemical

Catalog No. MSCU10KN100
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MSCU10KN100 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MSCU10KN100 Supplier Ricca Chemical Company Supplier No. MSCU10KN100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7440-50-8 , 7697-37-2 , 7732-18-5
Molecular Formula Cu
Molecular Weight (g/mol) 63.55
MDL Number MFCD00010965
InChI Key RYGMFSIKBFXOCR-UHFFFAOYSA-N
PubChem CID 23978
ChEBI CHEBI:30052
IUPAC Name copper
SMILES [Cu]

Specifications

Packaging Natural LDPE Bottle
Concentration or Composition Notes 10,000ppm Cu
CAS Min % 88.0, 8.94, 0.88
CAS Max % 92.0, 9.31, 0.92
Equivalence 1 mL = 10 mg Cu
Physical Form Liquid
Color Blue
pH < 2
Boiling Point Approximately 100°C
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Solubility Miscible with water
Solution Type Calibration Standard
Type ICP Single-element Standard Solution
Quantity 100 mL
Grade Laboratory
Chemical Name or Material Copper ICP-MS Standard
Show More Show Less