missing translation for 'onlineSavingsMsg'
Learn More

Copper(I) 2-Thiophenecarboxylate, TCI America™

Catalog No. C23121G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C23121G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C23121G Supplier TCI America Supplier No. C23121G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 68986-76-5
Molecular Formula C5H3CuO2S
Molecular Weight (g/mol) 190.683
MDL Number MFCD02183524
InChI Key SFJMFSWCBVEHBA-UHFFFAOYSA-M
Synonym copper i thiophene-2-carboxylate, cutc, unii-7519wbl07l, copper i thiophenecarboxylate, copper 2-thienylcarboxylate, cuprous 2-thienylcarboxylate, copper 2-thiophenecarboxylate, copper 1+ thiophene-2-carboxylate, copper i thiophene-2-carboxylate mi
PubChem CID 11194830
IUPAC Name copper(1+);thiophene-2-carboxylate
SMILES C1=CSC(=C1)C(=O)[O-].[Cu+]

Specifications

Quantity 1 g
Formula Weight 190.68
Physical Form Crystalline Powder
Chemical Name or Material Copper(I) 2-Thiophenecarboxylate