missing translation for 'onlineSavingsMsg'
Learn More

Copper AA Standard, Certified, 1000ppm ±5ppm (1mL = 1mg Cu), LabChem™

Catalog No. LC134007
Change view
Click to view available options
Quantity:
125 mL
500 mL
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC134007 125 mL Poly Bottle
LC134001 500 mL Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. LC134007 Supplier LabChem Supplier No. LC134007

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 19004-19-4 , 7697-37-2 , 7732-18-5

Specifications

Packaging Poly Bottle
CAS Max % 97.63, 2, 0.37
Identification Passes Test
Density 1g/mL
Physical Form Liquid
Concentration 1000 ppm ±5 ppm (1mL = 1mg Cu)
Linear Formula Cu(NO3)2·25H2O
UN Number UN3264
Color Blue/Green
Solubility Soluble in water
Quantity 125 mL
Formula Weight 232.59
Grade Certified
Chemical Name or Material Copper AA Standard, 1000ppm (1mL = 1mg Cu)
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
Harmful to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
Avoid release to the environment.
If in skin: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
Take off contaminated clothing and wash before reuse.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Dispose of contents/container to comply with local, state and federal regulations.
Recommended Storage Room Temperature