Learn More
Copan Diagnostics T-Shaped Spreaders
Catalog No. 23600896
Change view
Click to view available options
Quantity:
10/Peel Pk., 500 Spreaders/Box
Catalog No. Quantity
23600896 10/Peel Pk., 500 Spreaders/Box
1 options
Catalog No. 23600896 Supplier Copan Diagnostics Supplier No. COPTS10
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
T-shaped spreaders
T-Shaped Plastic Spreaders
  • Designed for spreading and dispersing liquids across the surface of agar culture plates
  • Used for performing bacterial CFU plate counts on water, milk, urine and other liquid samples
  • Smooth, rounded surfaces prevent any cutting or gouging of agar during inoculation of culture plates
  • T-Shape design means that even pressure is applied across the entire length of the T-bar in contact with the agar
  • Available individually sterile wrapped or in packs of five or ten pieces in a peel pack
Specifications
Shape T-Shape
For Use With (Equipment) Culture Plates
Quantity 10/Peel Pk., 500 Spreaders/Box
Material Plastic
Product Type Cell Spreader
Sterility Sterile
For Use With (Application) Designed for spreading and dispersing liquids across the surface of agar culture plates, used for performing bacterial CFU plate counts on water, milk, urine and other liquid samples