Learn More
Copan Diagnostics ESwab™ MRSA Collection System
Sample collection pack for various microbiology analyses
$243.69
Spécifications
| Sterility | Sterile |
|---|---|
| For Use With (Application) | For various microbiology analysis |
| Product Type | MRSA Collection System |
| Numéro de catalogue | Numéro du manufacturier. | Includes | Prix | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Numéro de catalogue | Numéro du manufacturier. | Includes | Prix | Quantity & Availability | |||||
23600903
|
Copan Diagnostics
CA493CE02 |
Pink Polypropylene screw-cap tube with internal conical shape filled with 1mL of Liquid Amies Medium, 1 pink regular size flock swab plus 1 white regular size flocked swab |
paquet de 50 chaque for $243.69
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
||||
Description
- Sterile
- Single use
- Pink Polypropylene screw-cap
- Applicator stick securely attaches to the cap after collection
- Tube with internal conical shape filled with 1mL of Liquid Amies Medium
- Sampling sites: Nose, throat, perineum
- 50 units per shelf pack
- 10 x 50 kits per box
Spécifications
| Sterile | |
| For various microbiology analysis | |
| MRSA Collection System |