missing translation for 'onlineSavingsMsg'
Learn More

COOMASSIE™ Brilliant Blue R-250, BAKER™, J.T. Baker™

Catalog No. 2002639
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
2002639 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 2002639 Supplier Avantor Supplier No. F79201
Only null left

A grade of chemicals for laboratory use

Chemical Identifiers

CAS 6104-59-2
Molecular Formula C45H44N3NaO7S2
Molecular Weight (g/mol) 825.97
MDL Number MFCD00041762
InChI Key HAEGGLCXLFWTBE-UHFFFAOYSA-M
Synonym C.I. 42660
PubChem CID 61365
IUPAC Name sodium {4-[(4-ethoxyphenyl)amino]phenyl}bis(4-{ethyl[(3-sulfonatophenyl)methyl]amino}phenyl)methylium
SMILES [Na+].CCOC1=CC=C(NC2=CC=C(C=C2)[C+](C2=CC=C(C=C2)N(CC)CC2=CC=CC(=C2)S([O-])(=O)=O)C2=CC=C(C=C2)N(CC)CC2=CC=CC(=C2)S([O-])(=O)=O)C=C1

Specifications

Formula Weight 825.97
Packaging Wide Mouth Amber Glass Bottle
Quantity 5 g
Chemical Name or Material COOMASSIE™ Brilliant Blue R-250