missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Extension Collar for CoolBox™ 2XT System

Catalog No. 07210090
Change view
Click to view available options
Color:
Green
Orange
Pink
Purple
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
07210090 Pink
07210087 Purple
07210088 Green
07210089 Orange
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options

Lid closure not possible with addition of extension collar

Catalog No. 07210090 Supplier Corning™ Supplier No. 432090

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Lid closure not possible with addition of extension collar

For use with CoolBox™ 2XT System. Corning™ Extension Collar is required when using taller CoolRack™ tube modules and tall tubes in CoolBox system.

For use with CoolBox XT and CoolBox 2XT to extend the height when using tall tube modules. The collar is magnetized and easily adheres to the unit base.

Specifications

Color Pink
For Use With (Application) Extend the height when using tall tube modules
For Use With (Equipment) CoolBox 2XT System
Quantity 1
Dimensions (L x W x H) Interior: 21.0 x 14.3 x 7.0 cm (8.2 x 5.6 x 2.7 in.)
Dimensions (L x W x H) Exterior 26.7 x 20.0 x 7.0 cm (6.5 x 7.8 x 2.7 in.)
Type Extension Collar