missing translation for 'onlineSavingsMsg'
Learn More

Supertek™ Convection Of Heat Apparatus

Catalog No. S012329
Encompass
Change view
Click to view available options
Class Size:
1 Lab Group
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S012329 1 Lab Group
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S012329 Supplier Supertek™ Supplier No. PH20320

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Designed for studying convection currents in the air, this apparatus consists of a metal box with glass front and two glass chimneys.

  • Designed for studying convection currents in the air.
  • Apparatus consists of a metal box, 230 x 75 x 75 mm, with glass front and two glass chimneys.
  • Supplied without candles

Specifications

Grade 6+
Class Size 1 Lab Group
Product Type Thermodynamic Apparatus