Learn More
Felix Storch Inc CONTROLLED RM TEMP ARS8PV-CRT
Numéro de catalogue. NC2256198
Encompass
ACCUCOLD ARS8PV-CRT (8cf) Controlled Room Temperature (CRT 68°F TO 77°F) cabinets are designed and purpose-built for ambient medication and vaccine storage in pharmacy and patient care areas. PRE-SET TO 72°F. Equipped with Hi/Lo Temp Alarm, Remote Contacts, open door alarm, power failure alarm, Sensor failure alarm, Self closing door, Hospital grade cord & plug, Access Port, Antimicrobial Handle. +/-1.5ºC Variation The published temperature variation is derived from the maximum deviation of an NTC sensor in a 1 oz. vial located nearest the chamber geometric center during a 24 hour test period. LED interior Light. Solid Door.
Numéro de catalogue. NC2256198 Fournisseur Felix Storch Inc Code fournisseur ARS8PVCRT
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
ACCUCOLD ARS8PV-CRT (8cf) Controlled Room Temperature (CRT 68°F TO 77°F) cabinets are designed and purpose-built for ambient medication and vaccine storage in pharmacy and patient care areas. PRE-SET TO 72°F. Equipped with Hi/Lo Temp Alarm, Remote Contacts, open door alarm, power failure alarm, Sensor failure alarm, Self closing door, Hospital grade cord & plug, Access Port, Antimicrobial Handle. +/-1.5ºC Variation The published temperature variation is derived from the maximum deviation of an NTC sensor in a 1 oz. vial located nearest the chamber geometric center during a 24 hour test period. LED interior Light. Solid Door.