missing translation for 'onlineSavingsMsg'
Learn More

Mopec™ Control Slides, IHC CD 20

Numéro de catalogue. 2244716
Change view
Click to view available options
Includes:
24 Unstained, 1 Stained
98 Unstained, 2 Stained
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Includes
2244716 24 Unstained, 1 Stained
2245249 98 Unstained, 2 Stained
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. 2244716 Fournisseur Mopec™ Code fournisseur SC08125
Il en reste null

All control slides are on Mopec™ IHC Enhanced Hydrophilic Adhesive Slides using Formalin Fixed Paraffin Embedded (FFPE) human tissue.

  • Product specific reactivity is guaranteed for one year from date of receipt when stored properly at 60°F to 80°F in a humidity controlled and light-deprived environment
  • Case of 25 contains 24 unstained slides and 1 stained slide
  • Case of 100 contains 98 unstained slides and 2 stained slides

Spécifications

Coating Uncoated
Thickness (Metric) 1 mm
Includes 24 Unstained, 1 Stained
Material Glass
Species Human
Tissue Condition Normal
Type IHC Control Slide
Dimensions (LxW) 75 x 25 mm
Enhanced Surface No